Benzylharnstoff, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
8.01795.0100EA
107
EUR
8.01795.0100
8.01795.9999
Benzylharnstoff, Sigma-Aldrich®
Benzylharnstoff
Formel:
C₈H₁₀N₂O Molecular Weight: 150,18 g/mol |
MDL:
MFCD00007951 CAS-Nummer: 538-32-9 EINECS: 208-689-1 |
Spezifikation Testergebnisse
Solubility | 17 g/l (45 °C) |
Melting Point | 143 - 146 °C |
Molar Mass | 150.17 g/mol |
Hill Formula | C8H10N2O |
Chemical Formula | C6H5CH2NHCONH2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...