(±)-2,3-Epoxypropylmethacrylat

Lieferant: POLYSCIENCES EUROPE
Achtung

POLS02607-500EA 0 EUR
POLS02607-500
(±)-2,3-Epoxypropylmethacrylat
(±)-2,3-Epoxypropylmethacrylat
Formel: C₇H₁₀O₃
Molecular Weight: 142,15 g/mol
MDL: MFCD00005137
CAS-Nummer: 106-91-2
EINECS: 203-441-9
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR