3-Hydroxypicolinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Hydroxypyridine-2-carboxylic acid

152307-25G 152307-5G
SIAL152307-25GEA 1170 EUR
SIAL152307-25G SIAL152307-5G
3-Hydroxypicolinsäure, Sigma-Aldrich®
3-Hydroxypicolinsäure
Formel: C₆H₅NO₃
Molecular Weight: 139,11 g/mol
MDL: MFCD00006294
CAS-Nummer: 874-24-8
EINECS: 212-859-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR