2-Bromisonicotinsäure, Sigma-Aldrich®

Lieferant: Merck

SIAL703990-5GEA 174 EUR
SIAL703990-5G SIAL703990-1G
2-Bromisonicotinsäure, Sigma-Aldrich®
2-Bromisonicotinsäure
Formel: C₆H₄BrNO₂
Molecular Weight: 202,01 g/mol
MDL: MFCD01646069
CAS-Nummer: 66572-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR