Thallium(I)chlorid ≥99,999% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Thalliumchlorid

11839.14
11839.14EA 294 EUR
11839.14
Thallium(I)chlorid ≥99,999% (Metall-Basis)
Thallium(I)chlorid

Purity: 99.999% CAS No.: 7791-12-0 Molecular Formula: TlCl Formula Weight: 239.82 Physical Form: Powder Density: 7.0 Refractive Index: 2.247 Sensitivity: Air Sensitive Melting Point: 430° Merck Reference: 14,9260 UN No.: 1707 MDL No.: MFCD00011274

Formel: ClTl
Molecular Weight: 239,84 g/mol
Siedepunkt: 720 °C (1013 hPa)
Schmelzpunkt: 430 °C
Dichte: 7,0
Lagertemperatur: Raumtemperatur
MDL: MFCD00011274
CAS-Nummer: 7791-12-0
EINECS: 232-241-4
UN: 1707
ADR: 6.1,II
Merck Index: 14,9260

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR