5-Bromisochinolin

Lieferant: Thermo Scientific
Achtung

CC05010DA CC05010.10
MAYBCC05010DAEA 0 EUR
MAYBCC05010DA CHMPCC05010.10
5-Bromisochinolin
5-Bromoisoquinoline
Formel: C₉H₆BrN
Molecular Weight: 208,06 g/mol
MDL: MFCD01646405
CAS-Nummer: 34784-04-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR