Hafnium(IV)chlorid ≥98% (Metall-Basis ausgeschlossen Zr) (max. 2,7% Zr)
Lieferant: Thermo Scientific
Gefahr
Synonyme: Hafnium tetrachloride
11834.14 11834.22 11834.88 AB202578.0025
11834.14EA 70.7 EUR
11834.14 11834.22 11834.88 ABCRAB202578.0025
Hafnium(IV)chlorid ≥98% (Metall-Basis ausgeschlossen Zr) (max. 2,7% Zr)
Hafnium(IV)chlorid
Purity: 98+% CAS No.: 13499-05-3 Molecular Formula: Cl4Hf Formula Weight: 320.30 Physical Form: Powder Sensitivity: Moisture Sensitive Appearance: White Odour: Irritating Melting Point: 320° subl. Merck Reference: 14,4588 UN No.: 3260 MDL No.: MFCD00011034
Formel: Cl₄Hf
Molecular Weight: 320,3 g/mol
Schmelzpunkt: 320 °C (Subl.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011034
CAS-Nummer: 13499-05-3
EINECS: 236-826-5
UN: 3260
ADR: 8,II
Merck Index: 14,4588