4-Aminoazobenzol

Lieferant: EHRENSTORFER GMBH

EHERC10167500EA 40 EUR
EHERC10167500
4-Aminoazobenzol
4-Aminoazobenzol
Formel: C₁₂H₁₁N₃
Molecular Weight: 197,24 g/mol
MDL: MFCD00007854
CAS-Nummer: 60-09-3
EINECS: 200-453-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR