4-Aminoazobenzol, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

46130-250MG-REA 44.6 EUR
46130-250MG-R
4-Aminoazobenzol, Supelco®
4-Aminoazobenzol
Formel: C₁₂H₁₁N₃
Molecular Weight: 197,24 g/mol
MDL: MFCD00007854
CAS-Nummer: 60-09-3
EINECS: 200-453-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR