Bis(2-ethylhexyl)phosphat ≥95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Bis(2-ethylhexyl) hydrogen phosphate, Di-(2-ethylhexyl)phosphoric acid
17723.22
17723.36
17723.A3
17723.22EA
36.4
EUR
17723.22
17723.36
17723.A3
Bis(2-ethylhexyl)phosphat ≥95%
Bis(2-ethylhexyl)phosphat
Purity: 95%
CAS No.: 298-07-7
Molecular Formula: C16H35O4P
Formula Weight: 322.43
Flash Point: 171°(340°F)
Physical Form: Liquid
Density: 0.965
Refractive Index: 1.4420
Appearance: Colourless
pH: 2-3
Melting Point: -50°
Boiling Point: 48°/12mm
UN No.: 1902
MDL No.: MFCD00009492
Extraction of metals
Formel:
C₁₆H₃₅O₄P Molecular Weight: 322,43 g/mol Siedepunkt: 48 °C (12 mmHg) Schmelzpunkt: –50 °C Dichte: 0,965 Flammpunkt: 171 °C (340 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009492 CAS-Nummer: 298-07-7 EINECS: 206-056-4 UN: 1902 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...