Bis(2-ethylhexyl)phosphat, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: Bis(2-ethylhexyl) hydrogen phosphate, Di-(2-ethylhexyl)phosphoric acid

M-Clarity™ quality level = MQ100

237825-100G 237825-25G 237825-500G
SIAL237825-100GEA 46.5 EUR
SIAL237825-100G SIAL237825-25G SIAL237825-500G
Bis(2-ethylhexyl)phosphat, Sigma-Aldrich®
Bis(2-ethylhexyl)phosphat
Formel: C₁₆H₃₅O₄P
Molecular Weight: 322,43 g/mol
MDL: MFCD00009492
CAS-Nummer: 298-07-7
EINECS: 206-056-4
UN: 1902
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR