Dibenzo[a,j]acridin

Lieferant: LGC Standards PROMOCHEM

PROOBCR-154EA 544 EUR
PROOBCR-154
Dibenzo[a,j]acridin
Dibenzo[a,j]acridin
Formel: C₂₁H₁₃N
CAS-Nummer: 224-42-0
UN: 1294
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR