Butyltrimethoxysilan

Lieferant: ABCR GMBH

ABCRAB110928.0100EA 0 EUR
ABCRAB110928.0100
Butyltrimethoxysilan
Butyltrimethoxysilan
Formel: C₇H₁₈O₃Si
Molecular Weight: 178,3 g/mol
CAS-Nummer: 1067-57-8
EINECS: 213-936-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR