Natrium-tert-butanolat ≥98%

Lieferant: ABCR GMBH
Gefahr

ABCRS931022.0100EA 0 EUR
ABCRS931022.0100
Natrium-tert-butanolat ≥98%
Natrium-tert-butanolat

Alkoxide

Formel: C₄H₉NaO
Molecular Weight: 96,1 g/mol
MDL: MFCD00040575
CAS-Nummer: 865-48-5
EINECS: 212-741-9
UN: 3206
ADR: 4.2,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR