L(+)-Ornithin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

SIAL75469-25GEA 66.3 EUR
SIAL75469-25G SIAL75469-500G
L(+)-Ornithin Hydrochlorid, Sigma-Aldrich®
L(+)-Ornithin Hydrochlorid
Formel: C₅H₁₂N₂O₂·HCl
Molecular Weight: 168,62 g/mol
MDL: MFCD00064562
CAS-Nummer: 3184-13-2
EINECS: 221-678-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR