Murexid-Indikator 0,25% in Natriumchlorid

Lieferant: Bernd Kraft

Synonyme: Ammonium-5-(2,4,6-trioxoperhydropyrimidin-5-ylidenamino)barbituratAmmoniumpurpuratPurpursäure Ammoniumsalz

KRAF06098.1600EA 102 EUR
KRAF06098.1600
Murexid-Indikator 0,25% in Natriumchlorid
Murexid
Formel: C₈H₈N₆O₆
Molecular Weight: 284,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012777
CAS-Nummer: 3051-09-0
EINECS: 221-266-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR