Fenclorim

Lieferant: EHRENSTORFER GMBH

EHERC13463000EA 80 EUR
EHERC13463000
Fenclorim
Fenclorim
Formel: C₁₀H₆Cl₂N₂
MDL: MFCD00754193
CAS-Nummer: 3740-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR