Fenclorim, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

46005-250MGEA 117 EUR
46005-250MG
Fenclorim, Supelco®
Fenclorim
Formel: C₁₀H₆Cl₂N₂
MDL: MFCD00754193
CAS-Nummer: 3740-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR