Catecholviolett

Lieferant: Thermo Scientific
Achtung

B23513.14EA 162 EUR
B23513.14 B23513.06
Catecholviolett
Catecholviolett
Formel: C₁₉H₁₄O₇S
Molecular Weight: 386,38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005868
CAS-Nummer: 115-41-3
EINECS: 204-088-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR