2,3,5,6-Tetrachlorphenol
Lieferant: LGC Standards PROMOCHEM
PROOCILULM24300.1EA
452
EUR
PROOCILULM24300.1
2,3,5,6-Tetrachlorphenol
2,3,5,6-Tetrachlorphenol
Formel:
C₆H₂Cl₄O Molecular Weight: 231,89 g/mol Siedepunkt: 261 °C (1013 hPa) Schmelzpunkt: 114…115 °C Dichte: 1,6 g/cm³ (20 °C) |
MDL:
MFCD00044671 CAS-Nummer: 935-95-5 EINECS: 213-310-8 UN: 2020 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...