Dibutylamin 0.2 mol/l (0.2 N) in DMF

Lieferant: Bernd Kraft
Gefahr

Synonyme: N-butyl-1-propanamin

KRAF10511.4700EA 341 EUR
KRAF10511.4700 KRAF10511.3700 KRAF10511.2700
Dibutylamin 0.2 mol/l (0.2 N) in DMF
Dibutylamin
Formel: C₈H₁₉N
Molecular Weight: 129,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009429
CAS-Nummer: 111-92-2
EINECS: 203-921-8
https://de.vwr.com/stibo/web/std.lang.all/03/15/2300315.jpg

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR