Terbufos (S-tert-Butylthiomethyl-O,O-diethyldithiophosphat)
Lieferant: EHRENSTORFER GMBH
EHERC17270000EA
58
EUR
EHERC17270000
Terbufos (S-tert-Butylthiomethyl-O,O-diethyldithiophosphat)
Terbufos (S-tert-Butylthiomethyl-O,O-diethyldithiophosphat)
Formel:
C₉H₂₁O₂PS₃ Molecular Weight: 288,44 g/mol |
CAS-Nummer:
13071-79-9 EINECS: 235-963-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...