Terbufos (S-tert-Butylthiomethyl-O,O-diethyldithiophosphat), Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

45313-100MGEA 79.1 EUR
45313-100MG
Terbufos (S-tert-Butylthiomethyl-O,O-diethyldithiophosphat), Supelco®
Terbufos (S-tert-Butylthiomethyl-O,O-diethyldithiophosphat)
Formel: C₉H₂₁O₂PS₃
Molecular Weight: 288,44 g/mol
CAS-Nummer: 13071-79-9
EINECS: 235-963-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR