Tetrakis(triphenylphosphin)palladium(0), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Pd(PPh3)4
SIAL10987-1GEA
103
EUR
SIAL10987-1G
SIAL10987-5G
Tetrakis(triphenylphosphin)palladium(0), Sigma-Aldrich®
Tetrakis(triphenylphosphin)palladium(0)
Formel:
C₇₂H₆₀P₄Pd Molecular Weight: 1155,58 g/mol Schmelzpunkt: 103…107 °C Dichte: 1 g/cm³ (20 °C) |
MDL:
MFCD00010012 CAS-Nummer: 14221-01-3 EINECS: 238-086-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...