Magnesiumcitrat

Lieferant: CAESAR & LORENTZ GMBH

CSLO2394-250GEA 36 EUR
CSLO2394-250G
Magnesiumcitrat
Magnesiumcitrat
Formel: C₆H₈O₇·nMg
MDL: MFCD00036422
CAS-Nummer: 7779-25-1
EINECS: 231-923-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR