Linuron (N-(3,4-Dichlorphenyl)-N'-methoxy,N'-methylharnstoff)

Lieferant: EHRENSTORFER GMBH

EHERC14640000EA 44 EUR
EHERC14640000
Linuron (N-(3,4-Dichlorphenyl)-N'-methoxy,N'-methylharnstoff)
Linuron (N-(3,4-Dichlorphenyl)-N'-methoxy,N'-methylharnstoff)
Formel: C₉H₁₀Cl₂N₂O₂
CAS-Nummer: 330-55-2
EINECS: 206-356-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR