Calciumcyanamid, Sigma-Aldrich®

Lieferant: Merck

SIAL438928-500GEA 124 EUR
SIAL438928-500G
Calciumcyanamid, Sigma-Aldrich®
Calcium cyanamide
Formel: CCaN₂
Molecular Weight: 80,11 g/mol
MDL: MFCD00064894
CAS-Nummer: 156-62-7
EINECS: 205-861-8
UN: 1403
ADR: 4.3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR