2-(Pentylthio)nicotinsäure

Lieferant: Thermo Scientific

BTB06369DA BTB06369EA BTB06369.50
MAYBBTB06369DAEA 0 EUR
MAYBBTB06369DA MAYBBTB06369EA MAYBBTB06369.50
2-(Pentylthio)nicotinsäure
2-(Pentylthio)nicotinsäure
Formel: C₁₁H₁₅NO₂S
CAS-Nummer: 175135-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR