3,6-Dimethyl-1,4-dioxan-2,5-dion

Lieferant: POLYSCIENCES EUROPE

POLS16640-100EA 0 EUR
POLS16640-100
3,6-Dimethyl-1,4-dioxan-2,5-dion
3,6-Dimethyl-1,4-dioxan-2,5-dion
Formel: C₆H₈O₄
Molecular Weight: 144,13 g/mol
MDL: MFCD00011685
CAS-Nummer: 95-96-5
EINECS: 202-468-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR