VWR®, Protease inhibitor cocktail, mammalian
Lieferant: VWR
VWR®
M250-1MLEA 105 EUR
M250-1ML
VWR®, Protease inhibitor cocktail, mammalian
Zellkultur-Ergänzungen und -Zusätze
Contains protease inhibitors for mammalian cell use: AEBSF, Aprotinin, E-64, Bestatin, Leupeptin and Pepstatin