Hygromycin B

Lieferant: STRATAGENE EUROPE
Gefahr

STRA200715EA 0 EUR
STRA200715
Hygromycin B
Hygromycin B
Formel: C₂₀H₃₇N₃O₁₃
Molecular Weight: 527,5 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06795479
CAS-Nummer: 31282-04-9
EINECS: 250-545-5
UN: 3462
ADR: 6.1,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR