Betain Monohydrat

Lieferant: Molekula

MOLE26904615-1KGEA 270 EUR
MOLE26904615-1KG
Betain Monohydrat
Betain Monohydrat
Formel: C₅H₁₁NO₂·1H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00150010
CAS-Nummer: 590-47-6
EINECS: 203-490-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR