Betain, Sigma-Aldrich®

Lieferant: Merck

Synonyme: TMG

M-Clarity™ quality level = MQ200

SIALB2629-1KGEA 166 EUR
SIALB2629-1KG SIALB2629-50G SIALB2629-100G
Betain, Sigma-Aldrich®
Betain
Formel: C₅H₁₁NO₂
Molecular Weight: 117,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012123
CAS-Nummer: 107-43-7
EINECS: 203-490-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR