Elektrodenhalter
Lieferant: Consort
CONSSH300EA 196 EUR
CONSSH300
Elektrodenhalter
Model SH300 holds up to three standard electrodes. It's heavy base and very stable flexible arm allow the electrodes to move sideways or up and down while keeping them at a constant vertical angle.