o-Kresolphthalein

Lieferant: Molekula

MOLE15685521-25GEA 105 EUR
MOLE15685521-25G
o-Kresolphthalein
o-Kresolphthalein
Formel: C₂₂H₁₈O₄
Molecular Weight: 346,39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005912
CAS-Nummer: 596-27-0
EINECS: 209-881-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR