Pluronic® F-68 Mn ~ 8,400, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Synperonic® F68
, Lutrol® F68
, PEG-PPG-PEG
, Kolliphor® P 188
, Pluronic® F-68
M-Clarity™ Qualitätsstufe = MQ100
SIAL412325-250GEA
99.8
EUR
SIAL412325-250G
Pluronic® F-68 Mn ~ 8,400, Sigma-Aldrich®
Kolliphor® P 188 (Lutrol® F68, Pluronic® F-68, Synperonic® F68)
Pluronic® F-68 LF Pastille. Hard and soft surface cleaners, defoamers in coatings and water treatment. Lubricant in metal working, anti-foaming aid and extender for linear and cross-linked polyesters and polyurethanes.
Formel:
HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a Molecular Weight: 8400 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082049 CAS-Nummer: 9003-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...