o-Kresolphthalein-Komplexon
Lieferant: Molekula
Synonyme:
Phthaleinpurpur
MOLEM82452467EA
0
EUR
MOLEM82452467
o-Kresolphthalein-Komplexon
o-Kresolphthalein-Komplexon
Formel:
C₃₂H₃₂N₂O₁₂ Molecular Weight: 636,62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005911 CAS-Nummer: 2411-89-4 EINECS: 219-318-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...