Creatin (N-Amidinosarkosin) Monohydrat 99%

Lieferant: Thermo Scientific

ACRO226791000EA 57.5 EUR
ACRO226791000 ACRO226795000 ACRO226790250
Creatin (N-Amidinosarkosin) Monohydrat 99%
Creatin (N-Amidinosarkosin) Monohydrat
Formel: C₄H₉N₃O₂·1H₂O
Molecular Weight: 149,15 g/mol (131,13 anhy)
Schmelzpunkt: 292 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00071582
CAS-Nummer: 6020-87-7
EINECS: 200-306-6

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤12.6 % (105°C)
Solubility (1 % in water) Clear colorless to light yellow
HPLC ≥98.5 % (on dry substance)
Residue after ignition ≤1 %
Titration with HClO4 ≥98.5 %
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR