Chromotrop 2R

Lieferant: Molekula

MOLE20169275-100GEA 191 EUR
MOLE20169275-100G
Chromotrop 2R
Chromotrop 2R
Formel: C₁₆H₁₀N₂Na₂O₈S₂
Molecular Weight: 468,37 g/mol
MDL: MFCD00003955
CAS-Nummer: 4197-07-3
EINECS: 224-085-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR