Puromycin Dihydrochlorid, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: (S)-2-Amino-N-((2S,3S,4R,5R)-5-(6-(Dimethylamino)-9H-purin-9-yl)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)-3-(4-methoxyphenyl)propanamide dihydrochloride

M-Clarity™ Qualitätsstufe = MQ300

P8833-100MG P8833-10MG P8833-25MG
SIALP8833-100MGEA 460 EUR
SIALP8833-100MG SIALP8833-10MG SIALP8833-25MG
Puromycin Dihydrochlorid, Sigma-Aldrich®
Puromycin Dihydrochlorid
Formel: C₂₂H₂₉N₇O₅·2HCl
Molecular Weight: 544,44 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00012691
CAS-Nummer: 58-58-2
EINECS: 200-387-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR