Adenosin freie Base ≥99%, hochrein

Lieferant: VWR Chemicals

0325-100GEA 294 EUR
0325-100G
Adenosin freie Base ≥99%, hochrein
Adenosin

A purine nucleoside essential for a variety of metabolic processes including energy transfer, signal transduction and mediation of inflammatory pathways.

Formel: C₁₀H₁₃N₅O₄
Molecular Weight: 267,24 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00005752
CAS-Nummer: 58-61-7
EINECS: 200-389-9

Bestellen

Spezifikation Testergebnisse

Em (257nm, 0.1N HCl, pH 2.0) ≥ 14600
Trocknungsverlust ≤ 0,5 %
Reinheit > 99 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR