Adenosin

Lieferant: Serva

SERA10770.02EA 0 EUR
SERA10770.02
Adenosin
Adenosin
Formel: C₁₀H₁₃N₅O₄
Molecular Weight: 267,24 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00005752
CAS-Nummer: 58-61-7
EINECS: 200-389-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR