Vancomycin Hydrochlorid
Lieferant: EHRENSTORFER GMBH
EHERC17900500EA
95
EUR
EHERC17900500
Vancomycin Hydrochlorid
Vancomycin Hydrochlorid
Formel:
C₆₆H₇₅Cl₂N₉O₂₄·HCl Molecular Weight: 1485,71 g/mol |
MDL:
MFCD03613611 CAS-Nummer: 1404-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...