ABTS (Diammonium-2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazol-6-sulfonat]), Sigma-Aldrich®
Lieferant: Merck Millipore (Oncogene)
Achtung
194430-1EA 122 EUR
194430-1
ABTS (Diammonium-2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazol-6-sulfonat]), Sigma-Aldrich®
ABTS (Diammonium-2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazol-6-sulfonat])
ABTS™ Chromophore, Diammonium Salt
Useful as a substrate for ELISA; acts as a hydrogen donor in peroxidase reactions. Radical cation has absorption maximum at 414 nm.
Formel: C₁₈H₂₄N₆O₆S₄
Molecular Weight: 548,68 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00010404
CAS-Nummer: 30931-67-0
EINECS: 250-396-6