ABTS (Diammonium-2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazol-6-sulfonat]), Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
SIAL11557-1GEA
104
EUR
SIAL11557-1G
ABTS (Diammonium-2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazol-6-sulfonat]), Sigma-Aldrich®
ABTS (Diammonium-2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazol-6-sulfonat])
Formel:
C₁₈H₂₄N₆O₆S₄ Molecular Weight: 548,68 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00010404 CAS-Nummer: 30931-67-0 EINECS: 250-396-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...