6-Methylycumarin

Lieferant: Thermo Scientific

Synonyme: 6-Methyl-2-benzopyron

MAYBSB00764.10EA 0 EUR
MAYBSB00764.10 MAYBSB00764.50 MAYBSB00764.1
6-Methylycumarin
6-Methylycumarin
Formel: C₁₀H₈O₂
Molecular Weight: 160,17 g/mol
Siedepunkt: 303 °C (1013 hPa)
Schmelzpunkt: 72…74 °C
MDL: MFCD00006875
CAS-Nummer: 92-48-8
EINECS: 202-158-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR