2,2',3,4',5',6-Hexachlorbiphenyl (PCB Nr. 149)
Lieferant: EHRENSTORFER GMBH
EHERC20014900EA
119
EUR
EHERC20014900
2,2',3,4',5',6-Hexachlorbiphenyl (PCB Nr. 149)
2,2',3,4',5',6-Hexachlorbiphenyl (PCB Nr. 149)
Formel:
C₁₂H₄Cl₆ Molecular Weight: 360,88 g/mol |
CAS-Nummer:
38380-04-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...