Ethosuximid

Lieferant: LGC Standards PROMOCHEM

PROOMM0750.00EA 235 EUR
PROOMM0750.00
Ethosuximid
Ethosuximid
Formel: C₇H₁₁NO₂
Molecular Weight: 141,17 g/mol
MDL: MFCD00072123
CAS-Nummer: 77-67-8
EINECS: 201-048-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR