Azidotrimethylsilan

Lieferant: ABCR GMBH

ABCRSIT8580.0.0050EA 0 EUR
ABCRSIT8580.0.0050 ABCRAB111513.0010
Azidotrimethylsilan
Azidotrimethylsilane
Formel: C₃H₉N₃Si
Molecular Weight: 115,21 g/mol
MDL: MFCD00001986
CAS-Nummer: 4648-54-8
EINECS: 225-078-5
UN: 1992
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR