1,3-Benzoxazol-6-amin

Lieferant: BIONET RESEARCH (CHEMPUR)

BIOT2P-046.5EA 0 EUR
BIOT2P-046.5 BIOT2P-046.10 BIOT2P-046.1 BIOT2P-046.500
1,3-Benzoxazol-6-amin
1,3-Benzoxazol-6-amin
Formel: C₇H₆N₂O
Molecular Weight: 134,10 g/mol
MDL: MFCD03791187
CAS-Nummer: 177492-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR